1,2-Dibromperfluorheptan 97%

Lieferant: Apollo Scientific

PC6844-25G PC6844-5G
APOSPC6844-25GEA 574 EUR
APOSPC6844-25G APOSPC6844-5G
1,2-Dibromperfluorheptan 97%
1,2-Dibromperfluorheptan
Formel: C₇Br₂F₁₄
Lagertemperatur: Raumtemperatur
MDL: MFCD03094409
CAS-Nummer: 678-32-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR