3-(Difluormethoxy)phenylacetonitril 98%

Lieferant: Apollo Scientific

PC6856-1G PC6856-5G PC6856-250MG
APOSPC6856-1GEA 0 EUR
APOSPC6856-1G APOSPC6856-5G APOSPC6856-250MG
3-(Difluormethoxy)phenylacetonitril 98%
3-(Difluormethoxy)phenylacetonitril
Formel: C₉H₇F₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00236241
CAS-Nummer: 41429-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR