1-(4-Fluoro-2-nitrophenyl)piperazine

Lieferant: Apollo Scientific
Achtung

PC6873-1G
APOSPC6873-1GEA 0 EUR
APOSPC6873-1G
1-(4-Fluoro-2-nitrophenyl)piperazine
1-(4-Fluoro-2-nitrophenyl)piperazine
Formel: C₁₀H₁₂FN₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00798371
CAS-Nummer: 243128-46-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR