2-Bromo-3-fluorobenzyl bromide 99%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-1-(bromomethyl)-3-fluorobenzene

PC6878-1G PC6878-25G PC6878-5G
APOSPC6878-1GEA 30.6 EUR
APOSPC6878-1G APOSPC6878-25G APOSPC6878-5G
2-Bromo-3-fluorobenzyl bromide 99%
2-Bromo-3-fluorobenzyl bromide
Formel: C₇H₅Br₂F
Lagertemperatur: Raumtemperatur
MDL: MFCD12911576
CAS-Nummer: 1184918-22-6
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR