3-Fluor-4-(trifluormethyl)benzophenon 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC6907-1G
APOSPC6907-1GEA
23.2
EUR
APOSPC6907-1G
3-Fluor-4-(trifluormethyl)benzophenon 97%
3-Fluor-4-(trifluormethyl)benzophenon
Formel:
C₁₄H₈F₄O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236282 CAS-Nummer: 243128-47-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...