3-Chlor-5-(trifluormethyl)benzonitril

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-Chloro-5-cyanobenzotrifluoride

PC6915-1G PC6915-25G PC6915-5G
APOSPC6915-1GEA 21.9 EUR
APOSPC6915-1G APOSPC6915-25G APOSPC6915-5G
3-Chlor-5-(trifluormethyl)benzonitril
3-Chlor-5-(trifluormethyl)benzonitril
Formel: C₈H₃ClF₃N
Molecular Weight: 205.57
Lagertemperatur: Raumtemperatur
MDL: MFCD04115854
CAS-Nummer: 693245-52-2
UN: 3276
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR