4-Bromo-2-(trifluoromethoxy)benzonitrile 99%

Lieferant: Apollo Scientific
Achtung

PC6918-1G PC6918-25G PC6918-5G
APOSPC6918-1GEA 0 EUR
APOSPC6918-1G APOSPC6918-25G APOSPC6918-5G
4-Bromo-2-(trifluoromethoxy)benzonitrile 99%
4-Bromo-2-(trifluoromethoxy)benzonitrile
Formel: C₈H₃BrF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD12761591
CAS-Nummer: 1187983-97-6
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR