2,5-Difluor-4-nitrophenol
Lieferant: Apollo Scientific
|
Gefahr
|
PC6921-1G
PC6921-5G
PC6921-250MG
APOSPC6921-1GEA
72.7
EUR
APOSPC6921-1G
APOSPC6921-5G
APOSPC6921-250MG
2,5-Difluor-4-nitrophenol
2,5-Difluor-4-nitrophenol
Formel:
C₆H₃F₂NO₃ Molecular Weight: 175.0897 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039297 CAS-Nummer: 120103-18-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...