Methyl 4-fluoro-2-methyl-5-nitrobenzoate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoro-5-(methoxycarbonyl)-4-methylnitrobenzene, 5-Fluoro-2-(methoxycarbonyl)-4-nitrotoluene

PC6924-1G
APOSPC6924-1GEA 21.9 EUR
APOSPC6924-1G
Methyl 4-fluoro-2-methyl-5-nitrobenzoate 98%
Methyl 4-fluoro-2-methyl-5-nitrobenzoate
Formel: C₉H₈FNO₄
Molecular Weight: 213,163 g/mol
Schmelzpunkt: 68…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD14581697
CAS-Nummer: 1163287-01-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR