4-Fluoro-2-methyl-5-nitrobenzoic acid 98%

Lieferant: Apollo Scientific
Achtung

PC6925-1G PC6925-5G
APOSPC6925-1GEA 21.9 EUR
APOSPC6925-1G APOSPC6925-5G
4-Fluoro-2-methyl-5-nitrobenzoic acid 98%
4-Fluoro-2-methyl-5-nitrobenzoic acid
Formel: C₈H₆FNO₄
Schmelzpunkt: 211…214 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11847453
CAS-Nummer: 64695-92-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR