4-(Dimethylamino)-4'-fluorochalcone

Lieferant: Apollo Scientific
Achtung

PC6932-1G
APOSPC6932-1GEA 0 EUR
APOSPC6932-1G
4-(Dimethylamino)-4'-fluorochalcone
4-(Dimethylamino)-4'-fluorochalcone
Formel: C₁₇H₁₆FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD00017963
CAS-Nummer: 28081-19-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR