1H,2H-Heptafluoro(1-iodo-3-methylbut-1-ene) 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC6932B-5G
APOSPC6932B-5GEA
63.8
EUR
APOSPC6932B-5G
1H,2H-Heptafluoro(1-iodo-3-methylbut-1-ene) 97%
1H,2H-Heptafluoro(1-iodo-3-methylbut-1-ene)
Formel:
C₅H₂F₇I Siedepunkt: 98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00155985 CAS-Nummer: 243139-61-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...