3,3',5,5'-Tetrakis(trifluormethyl)benzophenon 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC6934-1G
PC6934-5G
PC6934-10G
APOSPC6934-1GEA
0
EUR
APOSPC6934-1G
APOSPC6934-5G
APOSPC6934-10G
3,3',5,5'-Tetrakis(trifluormethyl)benzophenon 97%
3,3',5,5'-Tetrakis(trifluormethyl)benzophenon
Formel:
C₁₇H₆F₁₂O Schmelzpunkt: 139…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042474 CAS-Nummer: 175136-66-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...