Tetramethylammoniumhexafluorophosphat 99%

Lieferant: Apollo Scientific

Synonyme: Tetramethylammoniumhexafluorophosphate

PC6937-25G
APOSPC6937-25GEA 125 EUR
APOSPC6937-25G
Tetramethylammoniumhexafluorophosphat 99%
Tetramethylammonium hexafluorophosphate
Formel: C₄H₁₂F₆NP
Molecular Weight: 219,11 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012012
CAS-Nummer: 558-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR