N-(Oct-1-yl)-perfluoro-2,5-dimethyl-3,6-dioxanonanamide

Lieferant: Apollo Scientific

PC6939-5G
APOSPC6939-5GEA 236 EUR
APOSPC6939-5G
N-(Oct-1-yl)-perfluoro-2,5-dimethyl-3,6-dioxanonanamide
N-(Oct-1-yl)-perfluoro-2,5-dimethyl-3,6-dioxanonanamide
Formel: C₁₇H₁₈F₁₇NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00310125
CAS-Nummer: 340037-41-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR