Perfluoro-2,5-dimethyl-3,6-dioxanonanamide

Lieferant: Apollo Scientific

PC6940-1G PC6940-5G
APOSPC6940-1GEA 93.4 EUR
APOSPC6940-1G APOSPC6940-5G
Perfluoro-2,5-dimethyl-3,6-dioxanonanamide
Perfluoro-2,5-dimethyl-3,6-dioxanonanamide
Formel: C₉H₂F₁₇NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00347264
CAS-Nummer: 60308-67-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR