Tetrapropylammoniumtetrafluoroborat 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC6941-25G
APOSPC6941-25GEA
409
EUR
APOSPC6941-25G
Tetrapropylammoniumtetrafluoroborat 98%
Tetrapropylammonium tetrafluoroborate
Formel:
C₁₂H₂₈BF₄N Molecular Weight: 273,16 g/mol Schmelzpunkt: 245…247 °C Lagertemperatur: Raumtemperatur |
CAS-Nummer:
338-38-5 EINECS: 206-416-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...