2,3-Difluoro-4-(trifluoromethyl)benzaldehyde 95+%

Lieferant: Apollo Scientific
Achtung

PC6951-1G PC6951-5G PC6951-25G
APOSPC6951-1GEA 31.9 EUR
APOSPC6951-1G APOSPC6951-5G APOSPC6951-25G
2,3-Difluoro-4-(trifluoromethyl)benzaldehyde 95+%
2,3-Difluoro-4-(trifluoromethyl)benzaldehyde
Formel: C₈H₃F₅O
Lagertemperatur: Raumtemperatur
MDL: MFCD00236254
CAS-Nummer: 134099-20-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR