5-[3-Chloro-5-(trifluoromethyl)pyridin-2-ylsulphonyl]thiophene-2-sulphonyl chloride

Lieferant: Apollo Scientific

PC6954-100MG
APOSPC6954-100MGEA 288 EUR
APOSPC6954-100MG
5-[3-Chloro-5-(trifluoromethyl)pyridin-2-ylsulphonyl]thiophene-2-sulphonyl chloride
5-[3-Chloro-5-(trifluoromethyl)pyridin-2-ylsulphonyl]thiophene-2-sulphonyl chloride
Formel: C₁₀H₄Cl₂F₃NO₄S₃
Molecular Weight: 426,239 g/mol
Schmelzpunkt: 126…127 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00068006
CAS-Nummer: 175203-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR