5-Bromo-7-fluoroindole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-7-fluoro-1H-indole
PC6967-1G
APOSPC6967-1GEA
266
EUR
APOSPC6967-1G
5-Bromo-7-fluoroindole
5-Bromo-7-fluoroindole
Formel:
C₈H₅BrFN Molecular Weight: 214,04 g/mol Schmelzpunkt: 24…25 °C Flammpunkt: 110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04037876 CAS-Nummer: 883500-73-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...