Titan(IV)fluorid 99%
Lieferant: Apollo Scientific
|
Gefahr
|
PC6980-1G
APOSPC6980-1GEA
21.9
EUR
APOSPC6980-1G
Titan(IV)fluorid 99%
Titan(IV)fluorid
Formel:
F₄Ti Molecular Weight: 123,87 g/mol Schmelzpunkt: 248 °C (Subl.) Dichte: 2,798 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042545 CAS-Nummer: 7783-63-3 EINECS: 232-017-6 UN: 3260 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...