3-(Trifluormethoxy)toluol 98%

Lieferant: Apollo Scientific
Achtung

PC6982-25G PC6982-5G
APOSPC6982-25GEA 0 EUR
APOSPC6982-25G APOSPC6982-5G
3-(Trifluormethoxy)toluol 98%
3-(Trifluormethoxy)toluol
Formel: C₈H₇F₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD07366526
CAS-Nummer: 705-44-2
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR