2-Acetamido-4-fluorobenzeneboronic acid, pinacol ester 95%

Lieferant: Apollo Scientific

Synonyme: 2-(Acetylamino)-4-fluorobenzeneboronic acid, pinacol ester

PC6998-1G
APOSPC6998-1GEA 386 EUR
APOSPC6998-1G
2-Acetamido-4-fluorobenzeneboronic acid, pinacol ester 95%
2-Acetamido-4-fluorobenzeneboronic acid, pinacol ester
Formel: C₁₄H₁₉BFNO₃
Molecular Weight: 279,115 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12026077
CAS-Nummer: 1150271-67-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR