Allyl bromo(difluoro)acetate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Prop-2-en-1-yl 2-bromo-2,2-difluoroethanoate, Prop-2-en-1-yl bromo(difluoro)acetate

PC7004-1G
APOSPC7004-1GEA 292 EUR
APOSPC7004-1G
Allyl bromo(difluoro)acetate 95%
Allyl bromo(difluoro)acetate
Formel: C₅H₅BrF₂O₂
Molecular Weight: 214,993 g/mol
Siedepunkt: 30…32 °C (20 mm Hg)
Flammpunkt: 45 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08460495
CAS-Nummer: 155820-76-1
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR