4-Chlor-3-(trifluormethyl)phenylboronsäurepinakolester 97%

Lieferant: Apollo Scientific
Achtung

PC7018-1G PC7018-5G
APOSPC7018-1GEA 24.4 EUR
APOSPC7018-1G APOSPC7018-5G
4-Chlor-3-(trifluormethyl)phenylboronsäurepinakolester 97%
4-Chlor-3-(trifluormethyl)phenylboronsäurepinakolester
Formel: C₁₃H₁₅BClF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06657823
CAS-Nummer: 445303-09-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR