4-Chlor-3-(trifluormethyl)phenylboronsäurepinakolester 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-Chloro-3-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PC7018-5G
APOSPC7018-5GEA 56.7 EUR
APOSPC7018-5G
4-Chlor-3-(trifluormethyl)phenylboronsäurepinakolester 97%
4-Chlor-3-(trifluormethyl)phenylboronsäurepinakolester
Formel: C₁₃H₁₅BClF₃O₂
Molecular Weight: 306.516 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06657823
CAS-Nummer: 445303-09-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR