2,3-Difluor-4-methoxyphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Difluoro-4-methoxybenzeneboronic acid, (2,3-Difluoro-4-methoxyphenyl)boronic acid

PC7024-5G PC7024-25G
APOSPC7024-5GEA 71.2 EUR
APOSPC7024-5G APOSPC7024-25G
2,3-Difluor-4-methoxyphenylboronsäure
2,3-Difluor-4-methoxyphenylboronsäure
Formel: C₇H₇BF₂O₃
Lagertemperatur: Kühlschrank
MDL: MFCD07368234
CAS-Nummer: 170981-41-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR