2,6-Difluor-3-pyridinboronsäure Hydrat

Lieferant: Apollo Scientific
Achtung

PC7033-250MG
APOSPC7033-250MGEA 21.9 EUR
APOSPC7033-250MG
2,6-Difluor-3-pyridinboronsäure Hydrat
2,6-Difluor-3-pyridinboronsäure Hydrat
Formel: C₅H₄BF₂NO₂·H₂O
Lagertemperatur: Kühlschrank
MDL: MFCD06657881
CAS-Nummer: 1072952-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR