3-(Ethoxycarbonyl)-2-fluorphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Ethoxycarbonyl-2-fluorobenzeneboronic acid, 2-Fluoro-3-(ethoxycarbonyl)phenylboronic acid, (3-(Ethoxycarbonyl)-2-fluorophenyl)boronic acid

PC7035-100MG PC7035-1G PC7035-5G
APOSPC7035-100MGEA 21.9 EUR
APOSPC7035-100MG APOSPC7035-1G APOSPC7035-5G
3-(Ethoxycarbonyl)-2-fluorphenylboronsäure
3-(Ethoxycarbonyl)-2-fluorphenylboronsäure
Formel: C₉H₁₀BFO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD08064050
CAS-Nummer: 1072952-52-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR