2-Fluor-5-formylpyridin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Fluoropyridine-3-carboxaldehyde, 6-Fluoronicotinaldehyde

PC7037-1G PC7037-5G PC7037-10G
APOSPC7037-1GEA 26.1 EUR
APOSPC7037-1G APOSPC7037-5G APOSPC7037-10G
2-Fluor-5-formylpyridin 97%
2-Fluor-5-formylpyridin
Formel: C₆H₄FNO
Molecular Weight: 125.1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08064049
CAS-Nummer: 677728-92-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR