2-Fluor-3-methylphenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-Fluoro-3-methylphenyl)boronic acid
PC7045-1G
PC7045-5G
PC7045-25G
APOSPC7045-1GEA
21.9
EUR
APOSPC7045-1G
APOSPC7045-5G
APOSPC7045-25G
2-Fluor-3-methylphenylboronsäure
2-Fluor-3-methylphenylboronsäure
Formel:
C₇H₈BFO₂ Molecular Weight: 153.95 g/mol Schmelzpunkt: 77…79 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06409950 CAS-Nummer: 762287-58-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...