2-Fluor-6-methyl-3-pyridinboronsäurepinakolester 98%

Lieferant: Apollo Scientific

Synonyme: 2-Fluoro-6-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

PC7049-250MG
APOSPC7049-250MGEA 94.4 EUR
APOSPC7049-250MG
2-Fluor-6-methyl-3-pyridinboronsäurepinakolester 98%
2-Fluor-6-methyl-3-pyridinboronsäurepinakolester
Formel: C₁₂H₁₇BFNO₂
Molecular Weight: 237,078 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08063091
CAS-Nummer: 1218790-38-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR