3-Fluoro-2-(morpholin-4-yl)pyridine-4-boronic acid, pinacol ester

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)morpholine, 3-Fluoro-2-(morpholin-4-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 4-[3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]morpholine

PC7052-1G PC7052-5G
APOSPC7052-1GEA 181 EUR
APOSPC7052-1G APOSPC7052-5G
3-Fluoro-2-(morpholin-4-yl)pyridine-4-boronic acid, pinacol ester
3-Fluoro-2-(morpholin-4-yl)pyridine-4-boronic acid, pinacol ester
Formel: C₁₅H₂₂BFN₂O₃
Lagertemperatur: Kühlschrank
MDL: MFCD07781241
CAS-Nummer: 957198-29-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR