Kalium(2-fluor-4-(trifluormethyl)phenyl)trifluoroborat

Lieferant: Apollo Scientific

PC7058-1G PC7058-5G
APOSPC7058-1GEA 94.5 EUR
APOSPC7058-1G APOSPC7058-5G
Kalium(2-fluor-4-(trifluormethyl)phenyl)trifluoroborat
Potassium-2-fluoro-4-(trifluoromethyl)phenyltrifluoroborate
Formel: C₇H₃BF₇K
Lagertemperatur: Raumtemperatur
MDL: MFCD07781236
CAS-Nummer: 1150655-12-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR