Kalium(2,4-bis(trifluormethyl)phenyl)trifluoroborat

Lieferant: Apollo Scientific

PC7064-1G PC7064-5G
APOSPC7064-1GEA 154 EUR
APOSPC7064-1G APOSPC7064-5G
Kalium(2,4-bis(trifluormethyl)phenyl)trifluoroborat
Kalium(2,4-bis(trifluormethyl)phenyl)trifluoroborat
Formel: C₈H₃BF₉K
Lagertemperatur: Raumtemperatur
MDL: MFCD07368252
CAS-Nummer: 1150655-10-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR