(1R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethylamine hydrochloride 97%

Lieferant: Apollo Scientific

Synonyme: (R)-1-(3,5-Bis(trifluoromethyl)phenyl)ethanamine hydrochloride, (1R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanamine hydrochloride

PC7071-1G
APOSPC7071-1GEA 163 EUR
APOSPC7071-1G
(1R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethylamine hydrochloride 97%
(1R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethylamine hydrochloride
Formel: C₁₀H₉F₆N·ClH
Molecular Weight: 293,637 g/mol
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03426128
CAS-Nummer: 216002-20-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR