2-Fluor-1,3-dimethoxy-4-nitrobenzol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4-Dimethoxy-3-fluoronitrobenzene

PC7106-1G
APOSPC7106-1GEA 176 EUR
APOSPC7106-1G
2-Fluor-1,3-dimethoxy-4-nitrobenzol 98%
2-Fluor-1,3-dimethoxy-4-nitrobenzol
Formel: C₈H₈FNO₄
Molecular Weight: 201,1500 g/mol
Schmelzpunkt: 58…61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027197
CAS-Nummer: 155020-44-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR