5-Bromo-2-fluorophenylhydrazine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(5-Bromo-2-fluorophenyl)hydrazine hydrochloride, 4-Bromo-1-fluoro-2-hydrazinobenzene hydrochloride
PC7111-1G
APOSPC7111-1GEA
85.7
EUR
APOSPC7111-1G
5-Bromo-2-fluorophenylhydrazine hydrochloride
5-Bromo-2-fluorophenylhydrazine hydrochloride
Formel:
C₆H₆BrFN₂·ClH Molecular Weight: 241,489 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094411 CAS-Nummer: 214916-08-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...