2-Chloro-6-fluorophenylhydrazine hydrochloride 95%

Lieferant: Apollo Scientific

PC7115-1G
APOSPC7115-1GEA 48 EUR
APOSPC7115-1G
2-Chloro-6-fluorophenylhydrazine hydrochloride 95%
2-Chloro-6-fluorophenylhydrazine hydrochloride
Formel: C₆H₆ClFN₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD03094413
CAS-Nummer: 529512-79-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR