3-Fluoro-4-methylphenylhydrazine hydrochloride 95+%

Lieferant: Apollo Scientific

Synonyme: 2-Fluoro-4-hydrazinotoluene hydrochloride, (3-Fluoro-4-methylphenyl)hydrazine hydrochloride

PC7117-1G
APOSPC7117-1GEA 92.8 EUR
APOSPC7117-1G
3-Fluoro-4-methylphenylhydrazine hydrochloride 95+%
3-Fluoro-4-methylphenylhydrazine hydrochloride
Formel: C₇H₉FN₂·ClH
Molecular Weight: 176,619 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094415
CAS-Nummer: 156941-64-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR