N-Boc-3,5-difluoro-L-phenylalanine 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Boc-Phe(3,5-DiF)-OH
PC7119-1G
PC7119-250MG
PC7119-5G
APOSPC7119-1GEA
37.7
EUR
APOSPC7119-1G
APOSPC7119-250MG
APOSPC7119-5G
N-Boc-3,5-difluoro-L-phenylalanine 99%
N-Boc-3,5-difluoro-L-phenylalanine
Formel:
C₁₄H₁₇F₂NO₄ Schmelzpunkt: 115 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00797555 CAS-Nummer: 205445-52-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...