2-Fluoro-4-(methylsulphonyl)benzeneboronic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoro-4-(methylsulphonyl)phenylboronic acid

PC7122-250MG
APOSPC7122-250MGEA 98.9 EUR
APOSPC7122-250MG
2-Fluoro-4-(methylsulphonyl)benzeneboronic acid 98%
2-Fluoro-4-(methylsulphonyl)benzeneboronic acid
Formel: C₇H₈BFO₄S
Molecular Weight: 218.01 g/mol
Schmelzpunkt: 162…168 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027248
CAS-Nummer: 957060-85-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR