4-Borono-3-fluorthioanisol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoro-4-methylthiophenylboronic acid, (2-Fluoro-4-(methylthio)phenyl)boronic acid, 2-Fluoro-4-(methylsulphanyl)benzeneboronic acid

PC7124-250MG PC7124-500MG
APOSPC7124-250MGEA 87 EUR
APOSPC7124-250MG APOSPC7124-500MG
4-Borono-3-fluorthioanisol 98%
4-Borono-3-fluorthioanisol
Formel: C₇H₈BFO₂S
Molecular Weight: 186.0116 g/mol
Schmelzpunkt: 122…123 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09027247
CAS-Nummer: 957060-84-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR