5-(Trifluormethoxy)oxindol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Oxo-5-(trifluoromethoxy)indoline
PC7125-1G
APOSPC7125-1GEA
248
EUR
APOSPC7125-1G
5-(Trifluormethoxy)oxindol 98%
5-(Trifluormethoxy)oxindol
Formel:
C₉H₆F₃NO₂ Molecular Weight: 217,15 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09027275 CAS-Nummer: 403853-48-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...