2,3-Difluor-5-nitrophenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: (2,3-Difluoro-5-nitrophenyl)boronic acid

PC7127-1G
APOSPC7127-1GEA 176 EUR
APOSPC7127-1G
2,3-Difluor-5-nitrophenylboronsäure
2,3-Difluor-5-nitrophenylboronsäure
Formel: C₆H₄BF₂NO₄
Molecular Weight: 202,91 g/mol
Schmelzpunkt: 103…106 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09027242
CAS-Nummer: 957060-82-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR