2,3-Difluor-4-methoxybenzonitril 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Difluor-4-methoxybenzoesäurenitril

PC7137-1G
APOSPC7137-1GEA 65.4 EUR
APOSPC7137-1G
2,3-Difluor-4-methoxybenzonitril 98%
2,3-Difluor-4-methoxybenzonitril
Formel: C₈H₅F₂NO
Molecular Weight: 169,13 g/mol
Schmelzpunkt: 82…85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03428596
CAS-Nummer: 256417-12-6
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR