2'-Fluor-4'-iodacetanilid
Lieferant: Apollo Scientific
|
Achtung
|
PC7138-5G
PC7138-1G
PC7138-25G
APOSPC7138-5GEA
52.2
EUR
APOSPC7138-5G
APOSPC7138-1G
APOSPC7138-25G
2'-Fluor-4'-iodacetanilid
2'-Fluor-4'-iodacetanilid
Formel:
C₈H₇FINO Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039298 CAS-Nummer: 97760-94-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...