2,3-Dichlor-4-fluoranilin 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC7141-25G
PC7141-5G
APOSPC7141-25GEA
488
EUR
APOSPC7141-25G
APOSPC7141-5G
2,3-Dichlor-4-fluoranilin 95%
2,3-Dichlor-4-fluoranilin
Formel:
C₆H₄Cl₂FN Schmelzpunkt: 63…68 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08706409 CAS-Nummer: 36556-52-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...