1,2-Difluorpropan 97%

Lieferant: Apollo Scientific

Synonyme: FC-272ea

PC7149-5G
APOSPC7149-5GEA 541 EUR
APOSPC7149-5G
1,2-Difluorpropan 97%
1,2-Difluoropropane
Formel: C₃H₆F₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00040001
CAS-Nummer: 62126-90-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR