Chloro(fluoro)phenylacetonitrile 97%

Lieferant: Apollo Scientific
Achtung

PC7151-1G PC7151-250MG PC7151-5G
APOSPC7151-1GEA 261 EUR
APOSPC7151-1G APOSPC7151-250MG APOSPC7151-5G
Chloro(fluoro)phenylacetonitrile 97%
Chloro(fluoro)phenylacetonitrile
Formel: C₈H₅ClFN
Flammpunkt: >90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09258967
CAS-Nummer: 948014-31-1
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR